Anti-ACP5 Mouse Monoclonal Antibody [clone: 2D9]
Catalog # ABNOH00000054-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Acid Phosphatase 5
- Antigen symbol:ACP5
- Clonality:Monoclonal
- Clone:2D9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:54
- Antigen synonyms:SPENCDI|TRAP|HPAP
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ACP5
Clonality: Monoclonal
Clone: 2D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human