Anti-OTC Mouse Monoclonal Antibody [clone: OTI2C2]
Catalog # ORIGTA802595
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:ornithine carbamoyltransferase
- Antigen symbol:OTC
- Clonality:Monoclonal
- Clone:OTI2C2
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Amino acid number:33 to 354
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:36.1 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Protein
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: OTC (ornithine carbamoyltransferase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human