Anti-CRABP2 Mouse Monoclonal Antibody [clone: OTI3C11]
ORIGTA800027
: OriGene
- Antibody type:Primary
- Antigen name:Cellular retinoic acid binding Protein 2
- Antigen symbol:CRABP2
- Clonality:Monoclonal
- Clone:OTI3C11
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:15.5 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Protein
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:500-2000
Type: Primary
Antigen: CRABP2 (cellular retinoic acid binding protein 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human