Anti-ALDH1A3 Mouse Monoclonal Antibody [clone: OTI4A3]
Catalog # ORIGTA502840
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:aldehyde dehydrogenase 1 family, member A3
- Antigen symbol:ALDH1A3
- Clonality:Monoclonal
- Clone:OTI4A3
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Form:Liquid
- Amino acid number:1 to 100 and 413 to 512
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:55.9 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Protein
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150
Type: Primary
Antigen: ALDH1A3 (aldehyde dehydrogenase 1 family, member A3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human