Anti-GBA3 Mouse Monoclonal Antibody [clone: OTI4C7]
Catalog # ORIGTA502603
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:glucosidase, beta, acid 3 (gene/pseudogene)
- Antigen symbol:GBA3
- Clonality:Monoclonal
- Clone:OTI4C7
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Amino acid number:1 to 150 and 370 to 469
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:53.5 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Protein
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100
Type: Primary
Antigen: GBA3 (glucosidase, beta, acid 3 (gene/pseudogene))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human