Anti-SIGLEC9 Mouse Monoclonal Antibody [clone: OTI8F7]
Catalog # ORIGTA500380
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Sialic acid binding Ig-like lectin 9
- Antigen symbol:SIGLEC9
- Clonality:Monoclonal
- Clone:OTI8F7
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:50.1 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2500-5000; Immunochemistry: 1:50
Type: Primary
Antigen: SIGLEC9 (sialic acid binding Ig-like lectin 9)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: