Anti-SLC7A2 Rabbit Polyclonal Antibody
Catalog # ORIGTA317347
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 7 (cationic amino acid transporter, y+ system), member 2
- Antigen symbol:Solute carrier family 7, member 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Pk:50 µG
Specifications
About this item
- Recommended Dilutions: Immunochemistry: 15 µg/ml
Type: Primary
Antigen: SLC7A2 (solute carrier family 7 (cationic amino acid transporter, y+ system), member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human