Anti-SLC27A4 Rabbit Polyclonal Antibody
Catalog # ORIGTA315134
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 27 (fatty acid transporter), member 4
- Antigen symbol:Solute carrier family 27, member 4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Form:Liquid
- Storage buffer:Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:20000
Type: Primary
Antigen: SLC27A4 (solute carrier family 27 (fatty acid transporter), member 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse