Anti-OAT Rabbit Polyclonal Antibody
Catalog # ABGEAP8897A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Ornithine Aminotransferase
- Antigen symbol:OAT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:Center
- Gene ID:6645
- Antigen synonyms:D16S2531E|SNT2B2|EST25263|SNTL|SNT3
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:GSPKHQNSTKDRKIC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This SNTB2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 217-245 amino acids from the Central region of human SNTB2.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: OAT (ornithine aminotransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human