Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-AP2A2 Rabbit Polyclonal Antibody
  ABGEAP8551C
 :  Abgent
undefined
Anti-AP2A2 Rabbit Polyclonal Antibody
  ABGEAP8551C
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Adaptor-Related Protein Complex 2, Alpha 2 Subunit
  • Antigen symbol:
    AP2A2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-Terminal
  • Gene ID:
    5046
  • Antigen synonyms:
    KIAA0899|AP2A2|HYPJ|Adapter-related protein complex 2 alpha-2 subunit|AP-2 complex subunit alpha-2|Alpha2-adaptin|ADTAB|Alpha-adaptin C|HIP9|100 kDa coated vesicle protein C|Adaptor protein complex AP-2 subunit alpha-2|CLAPA2|Clathrin assembly
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Sequence:
    DEEDYTAQSTPGSANC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This PCSK6 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 659-688 amino acids from the C-terminal region of human PCSK6.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: AP2A2 (adaptor-related protein complex 2, alpha 2 subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human