Anti-NRAS Rabbit Polyclonal Antibody
ABGEAP7745B
: Abgent
- Antibody type:Primary
- Antigen name:Neuroblastoma RAS viral (v-ras) Oncogene homolog
- Antigen symbol:NRAS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Antigen synonyms:NRAS1|Neuroblastoma RAS|N-RAS|NS6|RASN|GTPase Nras|RAS|NRAS|ALPS4|Neuroblastoma RAS viral (v-ras) oncogene homolog|Transforming protein N-Ras
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: NRAS (neuroblastoma RAS viral (v-ras) oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human