Anti-OAZ1 Rabbit Polyclonal Antibody
Catalog # ABGEAP7477A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Ornithine decarboxylase antizyme 1
- Antigen symbol:OAZ1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:ODC-Az |OAZ|Ornithine decarboxylase antizyme
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: OAZ1 (ornithine decarboxylase antizyme 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse