Anti-RBKS (RBSK) Rabbit Polyclonal Antibody
Catalog # ABGEAP7182A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Ribokinase
- Antigen symbol:RBKS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-Terminal
- Cross adsorption:No
- Antigen synonyms:Ribokinase|RBKS
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: RBKS (RBSK)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human