Anti-MEF2C Rabbit Polyclonal Antibody
Catalog # ABGEAP6285C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Myocyte enhancer factor 2C
- Antigen symbol:MEF2C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:MADS box transcription enhancer factor 2 polypeptide C myocyte enhancer factor 2C|Myocyte enhancer factor 2C|Similar to MADS box transcription enhancer factor 2 polypeptide C|MEF2Cphospho S387|MEF2C_HUMAN.|Myocyte specic enhancer factor 2C
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: MEF2C (myocyte enhancer factor 2C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human