Anti-TERT Rabbit Polyclonal Antibody
Catalog # ABGEAP3544A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Telomerase Reverse Transcriptase
- Antigen symbol:TERT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:TRT|Telomerase catalytic subunit|Telomerase-associated protein 2|Telomerase reverse transcriptase|EST2|HEST2|TP2|TCS1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: TERT (telomerase reverse transcriptase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human