Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Anti-UTF1 Rabbit Polyclonal Antibody

Anti-UTF1 Rabbit Polyclonal Antibody

Catalog # ABGEAP17040C
Supplier:   Abgent
Anti-UTF1 Rabbit Polyclonal Antibody
Catalog # ABGEAP17040C
Supplier:  Abgent
Anti-UTF1 Rabbit Polyclonal Antibody
Anti-UTF1 Rabbit Polyclonal Antibody
Price
£312.00
Log in for availability
1 * 400 µl
Product Details & Documents

Type: Primary
Antigen: UTF1 (undifferentiated embryonic cell transcription factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human

Documents

Specifications
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Purification:
    Protein A & Peptide affinity purification
  • Antibody type:
    Primary
  • Epitope:
    Center
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Host:
    Rabbit
  • Sequence:
    RRYKFLKDKFREAHC
  • Concentration:
    0,5 mg/ml
  • Molecular weight:
    36.439 kDa
  • Shipping temperature:
    4 °C
  • Antigen name:
    Undifferentiated Embryonic Cell transcription Factor 1
  • Cat. No.:
    ABGEAP17040C
  • Reactivity:
    Human
  • Gene ID:
    8433
  • Antigen synonyms:
    UTF 1|hUTF 1|UTF1_HUMAN.|UTF1|hUTF1|Undferentiated embryonic cell transcription factor 1
  • Immunogen:
    This UTF1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 101-129 amino acids from the Central region of human UTF1.
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    UTF1
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • ELISA:
    Yes
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.