Anti-TLR6 Rabbit Polyclonal Antibody
Catalog # ABGEAP1506A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Toll-like Receptor 6
- Antigen symbol:TLR6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal ( 408–424 aa)
- Cross adsorption:No
- Antigen synonyms:CD antigen 286|CD286|Toll-like receptor 6|TLR6
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: TLR6 (toll-like receptor 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human