Anti-ENT1(Slc29a1) Rabbit Polyclonal Antibody
ABGEAP1086D
: Abgent
- Antibody type:Primary
- Antigen name:Solute Carrier Family 29 Member A1
- Antigen symbol:Solute carrier family 29, member 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal (491–506 aa)
- Cross adsorption:No
- Antigen synonyms:Solute carrier family 29 member 1|Equilibrative nucleoside transporter 1|ENT1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: ENT1(Slc29a1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human