Anti-OAZ2 Rabbit Polyclonal Antibody
Catalog # ABGEAP10765B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Ornithine decarboxylase antizyme 2
- Antigen symbol:OAZ2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:N-Terminal (1–100 aa)
- Cross adsorption:No
- Antigen synonyms:PFTK2|Serine/Threonine-Protein Kinase PFTAIRE-2|Amyotrophic Lateral Sclerosis 2 Chromosomal Region Candidate Gene 7 Protein|ALS2CR7
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: OAZ2 (ornithine decarboxylase antizyme 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse