Anti-ZNF763 Rabbit Polyclonal Antibody
Catalog # ABGEAP10563B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Zinc Finger Protein 763
- Antigen symbol:ZNF763
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal (648–660 aa)
- Cross adsorption:No
- Antigen synonyms:BARK1|BETA-ARK1|beta-adrenergic receptor kinase 1|G-protein coupled receptor kinase 2|GRK2|beta-ARK-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: ZNF763 (zinc finger protein 763)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human