Anti-USP54 Rabbit Polyclonal Antibody
AVIVARP69420_P050
- Antibody type:Primary
- Antigen name:ubiquitin specific peptidase 54
- Antigen symbol:USP54
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:159195
- Antigen synonyms:bA137L10.3|bA137L10.4|C10orf29
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: USP54 (ubiquitin specific peptidase 54)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human