Anti-DMKN Rabbit Polyclonal Antibody
AVIVARP66150_P050
- Antibody type:Primary
- Antigen name:dermokine
- Antigen symbol:DMKN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:93099
- Antigen synonyms:UNQ729|ZD52F10
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: DMKN (dermokine)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human