Anti-SPG21 Rabbit Polyclonal Antibody
Catalog # AVIVARP65713_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:spastic paraplegia 21 (autosomal recessive, Mast syndrome)
- Antigen symbol:SPG21
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:51324
- Antigen synonyms:GL010|ACP33|MAST|BM-019
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SPG21 (spastic paraplegia 21 (autosomal recessive, Mast syndrome))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human