Anti-TNKS1BP1 Rabbit Polyclonal Antibody
Catalog # AVIVARP58347_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Tankyrase 1 Binding Protein 1
- Antigen symbol:TNKS1BP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:85456
- Antigen synonyms:TAB182
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TNKS1BP1 (tankyrase 1 binding protein 1, 182kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human