Order Entry
United Kingdom
ContactUsLinkComponent
Anti-ISYNA1 Rabbit Polyclonal Antibody
Anti-ISYNA1 Rabbit Polyclonal Antibody
Catalog # AVIVARP53716_P050
undefined
Anti-ISYNA1 Rabbit Polyclonal Antibody
Catalog # AVIVARP53716_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Inositol-3-phosphate Synthase 1
  • Antigen symbol:
    ISYNA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    51477
  • Antigen synonyms:
    INOS|INO1|IPS|IPS-1|IPS 1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: ISYNA1 (inositol-3-phosphate synthase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: