Anti-OAZ3 Rabbit Polyclonal Antibody
AVIVARP53713_P050
- Antibody type:Primary
- Antigen name:ornithine decarboxylase antizyme 3
- Antigen symbol:OAZ3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:51686
- Antigen synonyms:AZ3|OAZ-t|TISP15
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: OAZ3 (ornithine decarboxylase antizyme 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: