Anti-CREG2 Rabbit Polyclonal Antibody
AVIVARP53207_P050
- Antibody type:Primary
- Antigen name:Cellular repressor of E1A-stimulated genes 2
- Antigen symbol:CREG2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Pig,Human,Bovine,Guinea pig,Rat,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:200407
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: CREG2 (cellular repressor of E1A-stimulated genes 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Guinea pig, Human, Mouse, Pig, Rat