Anti-UNQ1887 Rabbit Polyclonal Antibody
AVIVARP52668_P050
- Antibody type:Primary
- Antigen name:Signal Peptide Peptidase like 3
- Antigen symbol:SPPL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:121665
- Antigen synonyms:MDHV1887|IMP2|PSH1|PRO4332|PSL4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: UNQ1887
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human