Anti-CHAD Rabbit Polyclonal Antibody
AVIVARP51925_P050
- Antibody type:Primary
- Antigen name:Chondroadherin
- Antigen symbol:CHAD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:1101
- Antigen synonyms:SLRR4A
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: CHAD (chondroadherin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: