Anti-CCDC19 Rabbit Polyclonal Antibody
Catalog # AVIVARP51321_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Cilia and flagella associated protein 45
- Antigen symbol:CFAP45
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:25790
- Antigen synonyms:NESG1|CCDC19
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CCDC19 (coiled-coil domain containing 19)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: