Anti-OXSR1 Rabbit Polyclonal Antibody
Catalog # AVIVARP48661_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:oxidative-stress responsive 1
- Antigen symbol:OXSR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:108737
- Antigen synonyms:2210022N24Rik|AI462649|AW209236|Osr1|2810422B09Rik|mKIAA1101
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: OXSR1 (oxidative stress responsive 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: