Anti-OAT Rabbit Polyclonal Antibody
AVIVARP48135_T100
- Antibody type:Primary
- Antigen name:ornithine aminotransferase
- Antigen symbol:OAT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:4942
- Antigen synonyms:OATASE|OKT|HOGA|GACR
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Type: Primary
Antigen: OAT (ornithine aminotransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: