Anti-LUC7L3 Rabbit Polyclonal Antibody
AVIVARP40246_P050
- Antibody type:Primary
- Antigen name:LUC7-like 3
- Antigen symbol:LUC7L3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: LUC7L3 (LUC7-like 3 (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: