Anti-RNF113B Rabbit Polyclonal Antibody
Catalog # AVIVARP40028_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Ring Finger Protein 113B
- Antigen symbol:RNF113B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:140432
- Antigen synonyms:bA10G5.1|RNF161|ZNF183L1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: RNF113B (ring finger protein 113B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human