Anti-GATAD1 Rabbit Polyclonal Antibody
AVIVARP33193_P050
- Antibody type:Primary
- Antigen name:GATA zinc finger domain containing 1
- Antigen symbol:GATAD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:57798
- Antigen synonyms:ODAG|RG083M05.2|CMD2B
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: GATAD1 (GATA zinc finger domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: