Anti-AAV5 VP1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-10180R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Adeno-associated virus 5 capsid Protein VP1
- Antigen symbol:AAV5 VP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Virus
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Antigen synonyms:Capsid protein VP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: AAV5 VP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus