SPE columns, ISOLUTE® PAH HC
Catalog # ARGO910-0100-C
Supplier: Argonaut Technologies
Specifications
- Capacity:6 ml
- Seq:01
- Bed weight:1000 mg
- Column hardware:PP
- Pk:30
Specifications
About this item
This is an SPE column developed to rapidly clean up hexane extracts of PAHs from oil contaminated soils. The column is designed to separate the aromatic PAHs from aliphatic hydrocarbon interferences (up to C30), giving a clean baseline in GC analysis.