Whole sheep blood
Supplier: Rockland Immunochemicals
Certificates
About this item
Blood is obtained from healthy, non-medicated animals. For defibrinated blood, fibrin removed mechanically during the clotting process without the addition of anticoagulants. Defibrinated sheep blood is ideal for blood agar plate preparations.
Blood in Alsever's and sodium citrate is used for titration of complement, adsorption procedures, testing for agglutinins/HA assays and for the preparation of stroma as particulate reagents.
Frequently Bought Together