Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Halogen lamps
Halogen fibre optic lamp, with smooth reflector finish, Dental, microscope and medical applications, Osram 64255, G4, 8 V, 20 W
  ACEA64255
Halogen lamps
  ACEA64255
Halogen lamps
Halogen lamps
Halogen fibre optic lamp, with smooth reflector finish, Dental, microscope and medical applications, Osram 64255, G4, 8 V, 20 W
£37.90
1 * 1 items
  • For:
    Dental, microscope and medical applications
  • Description:
    Halogen fibre optic lamp, with smooth reflector finish
  • Seq:
    08
  • Base:
    G4
  • Model:
    Osram 64255
  • Power:
    20 W
  • Voltage:
    8 V