Order Entry
United Kingdom
ContactUsLinkComponent
Racks for LTS® tips (Rainin)
Racks for LTS® tips (Rainin)
Catalog # RAIN30389354
Supplier:  RAININ
Racks for LTS® tips (Rainin)
Catalog # RAIN30389354
Supplier:  RAININ

Specifications

  • Colour code:
    Red
  • Description:
    Green-Pak® rack, empty, for Green-Pak® individual and SpaceSaver® refills
  • Seq:
    01
  • For use with:
    10 μl refills
  • Pk:
    10

Specifications

About this item