Storage drums
5 L, 198×148×240 mm
Catalog # MEDL202605
Supplier: MEDLINE SCIENTIFIC
Jump to:
Product Details & Documents
Stackable drum, HDPE, with tamper evident screw cap. The four flat sides of the drum are suitable for sticking information labels. It is ideal for transport of hazardous and toxic substances.
- Good chemical resistance
- Good temperature resistance
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
- Capacity:5 L
- W×D×H:198×148×240 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



