General purpose thermometers
These stem type thermometers with red filing are suitable for general purpose applications.
- Permanent graduation and lettering
- With loop
- Capillary diameter: 6 ±1 mm
Frequently Bought Together
These stem type thermometers with red filing are suitable for general purpose applications.