Order Entry
United Kingdom
ContactUsLinkComponent
Suface and instrument cleaner, Umonium38®
Suface and instrument cleaner, Umonium38®
Catalog # LOCAPF12105-00
Supplier:  Local
undefined
Suface and instrument cleaner, Umonium38®
Catalog # LOCAPF12105-00
Supplier:  Local
Specifications
  • Description:
    Umonium38®, Surface, instrument and equipment cleaner
  • Environmentally Preferable:
  • Packed:
    1 l dispenser
  • Pk:
    12 L
Specifications
About this item
Umonium 38® Instruments is designed for the cleaning and disinfection of instruments and heat-sensitive and heat-resistant medical equipment. It can be used for manual washing of the instriments or cleaning in an ultrosonic bath.
  • Aldehyde-free
  • Efficient, fast and efficient (dissolves blood)
  • Good material compatibility and no residue after rinsing
  • Non corrosive, pH 7
  • Broad spectrum of activity: Mycobactericidal, bactericidal, fungicidal and virucidal
Certifications: Bactericide EN 1040, EN 13727; Fungicide EN 14561, EN 1275, EN 13624, EN 14562; Mycobactericide EN 14348, EN 14563; Virucide EN 14476.