Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Sub blotting system, omniPAGE
Sub blotting system, omniPAGE
Supplier:   CLEAVER SCIENTIFIC
Sub blotting system, omniPAGE
Supplier:  CLEAVER SCIENTIFIC
Certificates
Sub blotting system, omniPAGE
3 Options Available Below
Product Details & Documents
Each omniPAGE system comprises an omniPAGE tank and lid, electroblotting insert, cassettes and fibre pads. The versatile design of the omniPAGE tank and lid allows the unit to be easily adapted for vertical PAGE or IEF with capillary tube gels, using the relevant optional insert.
  • Multi-transfer capability for 4 mini gels
  • Complete run-time flexibility from overnight to as short as a couple of hours
  • Cool pack for environmentally friendly low-cost cooling during high-intensity transfers
  • Open design, rigid cassettes maximise current transfer and eliminate shadow band formation
  • Cassettes colour coded prevent polarity reversal to ensure transfer in the correct orientation
Specifications for Product Family
Specifications
  • Catalog No:
  • CLEVSB10
  • CLEVSB10W
  • CLEVSB20
  • Buffer volume:
  • 1200 ml
  • 2800 ml
  • 5600 ml
  • Gel size W×L:
  • 80×85 mm
  • 160×85 mm
  • 160×175 mm
Specifications
Product Family Options
Product Information
  • Dimensions
    Availability
    Price
  • CLEVSB10
    Mini blotting system
    Mini blotting system
    100×100 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - £1,100.00
    £1,100.00
    1 * 1 items
       £1,100.00
    Specifications:
    More
    Specifications
    Cat. No.
    CLEVSB10
    Buffer volume
    1200 ml
    Gel size W×L
    80×85 mm
  • CLEVSB10W
    Mini-wide blotting system
    Mini-wide blotting system
    100×200 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - £1,420.00
    £1,420.00
    1 * 1 items
       £1,420.00
    Specifications:
    More
    Specifications
    Cat. No.
    CLEVSB10W
    Buffer volume
    2800 ml
    Gel size W×L
    160×85 mm
  • CLEVSB20
    Maxi blotting system
    Maxi blotting system
    200×200 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - £1,730.00
    £1,730.00
    1 * 1 items
       £1,730.00
    Specifications:
    More
    Specifications
    Cat. No.
    CLEVSB20
    Buffer volume
    5600 ml
    Gel size W×L
    160×175 mm
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Fibre pads for 20×10 cm mini-wide blotter
    Fibre pads for 20×10 cm mini-wide blotter
    CLEVSB10WF
    Log in for availability
    1 * 6 items - £86.60
Product Information
  • Catalog #
    Availability
    Price
  • Compression cassette for 20×10 cm mini-wide blotter
    Compression cassette for 20×10 cm mini-wide blotter
    CLEVSB10WC
    Log in for availability
    1 * 1 items - £86.60
Product Information
  • Catalog #
    Availability
    Price
  • Mini-wide electroblotting module
    Mini-wide electroblotting module
    CLEVVS10WBI
    Log in for availability
    1 * 1 items - £754.00
Product Information
  • Catalog #
    Availability
    Price
  • Fibre pads for 20×20 cm maxi blotter
    Fibre pads for 20×20 cm maxi blotter
    CLEVSB20F
    Log in for availability
    1 * 4 items - £122.00
Product Information
  • Catalog #
    Availability
    Price
  • Maxi electroblotting module
    Maxi electroblotting module
    CLEVVS20BI
    Log in for availability
    1 * 1 items - £985.00
Product Information
  • Catalog #
    Availability
    Price
  • Fibre pads for 10×10 cm mini blotter
    Fibre pads for 10×10 cm mini blotter
    CLEVSB10F
    Log in for availability
    1 * 6 items - £72.70
Product Information
  • Catalog #
    Availability
    Price
  • Mini internal electroblotting module
    Mini internal electroblotting module
    CLEVVS10BI
    Log in for availability
    1 * 1 items - £661.00
Product Information
  • Catalog #
    Availability
    Price
  • Compression cassette for 20×20 cm maxi blotter
    Compression cassette for 20×20 cm maxi blotter
    CLEVSB20C
    Log in for availability
    1 * 1 items - £122.00
Product Information
  • Catalog #
    Availability
    Price
  • Compression cassette for 10×10 cm mini blotter
    Compression cassette for 10×10 cm mini blotter
    CLEVSB10C
    Log in for availability
    1 * 1 items - £72.70