Deep well plates, 96 square well, Biomek™
Catalog # BECL140504
Supplier: Beckman Coulter
Specifications
- Packed:24 plates/case
- Sterile:-
- Colour:Natural
- Description:Deep well plate, 96-well, PP, 2 ml, square well
- Seq:01
- Pk:24
- Well volume:2 ml
Specifications
About this item
PP. Biomek deep square 96-well plates are ideal for sample collection, compound preparation, combinatorial chemistry, high-throughput screening, nucleic acid purification, bacterial culture growth and plate replication.
- Automation-friendly
- Ideal for use with Biomek™ cap mats and foil lids for short- and long-term storage