Order Entry
United Kingdom
ContactUsLinkComponent
Hand disinfectants, Spirigel® Complete
Hand disinfectants, Spirigel® Complete
Catalog # SHIE3077760
Hand disinfectants, Spirigel® Complete
Catalog # SHIE3077760
Specifications
  • Description:
    Hand disinfectant, Spirigel® Complete, pump bottles, 12×500 ml
  • Seq:
    02
  • Environmentally Preferable:
  • Pk:
    12
Specifications
About this item
Spirigel® Complete hand sanitiser is a validated alcohol gel suitable for all areas. It is ideal to quickly and effectively decontaminate hands without water.
  • Bactericidal, virucidal, yeasticidal and mycobactericidal
  • Effective against Norovirus and Rotavirus in 15 seconds according to EN14476
  • Contains glycerol as emollient
  • Low-viscosity formulation with emollients for excellent skin compatibility
  • Clear, colourless and perfume-free
  • Dermatologically tested