Stainless steel cleaner, Suma Inox D7.1, Diversey
BUNZ042504
: BUNZL
- Description:Suma Inox D7.1, stainless steel cleaner,
- Packed:750 ml bottle with trigger spray
- Pk:6
Suma Inox D7.1 is a ready to use one step light duty cleaner/polish for use on stainless steel surfaces that are not used in the preparation of food. It gives shine back to stainless steel surfaces, removes spots and marks, and resists finger marks to maintain the just polished appearance for longer.