Airborne disinfectants, Airstel
Supplier: TRISTEL SOLUTIONS
Certificates
About this item
Developed for rapid decontamination of inaccessible surfaces and to reduce cross infection in high risk environments. It does not contain flammable gases and can be used in areas such as high-risk zones, chemical industry applications, operating rooms, aeroplanes and ambulances.
- Decontamination of microorganisms
- Effective elimination of unpleasant odours
- Environmental and surface disinfection
- Total discharge valve allows dispersing of contents in a single application
Caution: Use biocides safely. Always read the label and product information before use.
Frequently Bought Together