Detergent, DEC-CYCLE®
: PMT
DEC-CYCLE® is a phosphate-free germicidal detergent which is used to clean, disinfect, and deodorise any washable inanimate surface.
- Available in gamma irradiated unit dose bottles or in non sterile, 1 gallon containers
- Filtered at 0,2 µm
- Typical dosage level: 15 ml per 3,78 litres (US: 0,5 oz/gallon)
- Effective against a broad spectrum of pathogenic bacteria
It is a low pH, phenolic, dilutable, hospital detergent effective in 10 minutes at 20 °C in hard water up to 400 ppm (calculated as CaCO₃) in the presence of 5% blood serum. It is recommended for use in pharmaceutical, biotechnology, medical device manufacturing, hospitals and any health care institutions.
: The case of sterile unit dose bottles is supplied with lot specific Certificate of Irradiation, Certificate of Analysis, LAL Testing Analysis and Sterility Report. The non sterile version is supplied with certificate of conformance. Please contact Avantor for more application information and documentation details.
Frequently Bought Together