Order Entry
United Kingdom
ContactUsLinkComponent
Polish remover, Cleanline Easy Strip
Polish remover, Cleanline Easy Strip
Catalog # BUNZ051422
Supplier:  BUNZL
undefined
Polish remover, Cleanline Easy Strip
Catalog # BUNZ051422
Supplier:  BUNZL
Specifications
  • Capacity:
    5 l
  • Description:
    Cleanline Easy Strip polish remover
  • Pk:
    5 L
Specifications
About this item
Cleanline Easy strip remover is an alternative to the traditional caustic based polish strippers. It is suitable for use in well ventillated areas.